Learn More
Apexbio Technology LLC Enoxacin (Penetrex) 74011-58-8 50mg
SDP
Catalog No. 502240580
Encompass_Preferred
Enoxacin (Penetrex) (CAS 74011-58-8) is a small-molecule inhibitor targeting bacterial DNA gyrase and topoisomerase IV It is designed to interfere with bacterial DNA replication and transcription by inhibiting these essential enzymes thereby disrupting cellular DNA processes Enoxacin exerts its biological activity primarily through stabilization of enzyme-DNA cleavage complexes leading to inhibition of DNA replication In in vitro studies enoxacin demonstrates inhibitory activity with reported IC50 values against bacterial DNA gyrase typically ranging between 0 5 10 g/mL depending on bacterial species and assay conditions Based on these pharmacological properties enoxacin holds research potential in studies of DNA topology bacterial resistance mechanisms and antimicrobial pharmacodynamics
This item is not returnable. View return policy
Catalog No. 50-224-0580 Supplier Apexbio Technology LLC Supplier No. A842350
Enoxacin (Penetrex) (CAS 74011-58-8) is a small-molecule inhibitor targeting bacterial DNA gyrase and topoisomerase IV It is designed to interfere with bacterial DNA replication and transcription by inhibiting these essential enzymes thereby disrupting cellular DNA processes Enoxacin exerts its biological activity primarily through stabilization of enzyme-DNA cleavage complexes leading to inhibition of DNA replication In in vitro studies enoxacin demonstrates inhibitory activity with reported IC50 values against bacterial DNA gyrase typically ranging between 0 5 10 g/mL depending on bacterial species and assay conditions Based on these pharmacological properties enoxacin holds research potential in studies of DNA topology bacterial resistance mechanisms and antimicrobial pharmacodynamics