Learn More
Selleck Chemical LLC Enoxacin Sesquihydrate S5209-25mg
SDP
Catalog No. 503112757
Encompass_Preferred
Enoxacin sesquihydrate (Enoxacin hydrate) is a broad-spectrum 6-fluoronaphthyridinone antibacterial agent
Catalog No. 50-311-2757 Supplier Selleck Chemical LLC Supplier No. S520925MG
Enoxacin sesquihydrate (Enoxacin hydrate) is a broad-spectrum 6-fluoronaphthyridinone antibacterial agent