Learn More
Selleck Chemical LLC Enoxolone 500mg 471-53-4 Glycyrrhetin
SDP
Catalog No. 505836
Encompass_Preferred
Enoxolone (Glycyrrhetin) is a pentacyclic triterpenoid derivative of the beta-amyrin type obtained from the hydrolysis of glycyrrhizic acid, which was obtained from the herb liquorice. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-583-6 Supplier Selleck Chemical LLC Supplier No. S2296500MG
Enoxolone (Glycyrrhetin) is a pentacyclic triterpenoid derivative of the beta-amyrin type obtained from the hydrolysis of glycyrrhizic acid, which was obtained from the herb liquorice. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.