Learn More
Medchemexpress LLC Enozertinib hemihydrate | 2489185-38-6 | 100.0% | 669.77 | C35H42F2N8O3·1/2H2O | 10 MG
SDP
Catalog No. 5000295420
Encompass_Preferred
Enozertinib hemihydrate is a small-molecule, brain-penetrant inhibitor of EGFR and HER2 supplied as a hemihydrate for research use. It is orally bioavailable and exhibits antineoplastic activity against EGFR-altered tumors, including exon 20 insertions; intended for laboratory research and not for human use.
  • Hemihydrate form for consistent handling and stability.
  • High purity suitable for biochemical and cellular assays.
  • Brain-penetrant and orally bioavailable properties.
  • Targets EGFR and HER2 alterations, including exon 20 insertions.
  • Supplied in small-mass packs for preclinical research.
Catalog No. 50-002-95420 Supplier Medchemexpress LLC Supplier No. HY172429A10MG
Enozertinib hemihydrate is a small-molecule, brain-penetrant inhibitor of EGFR and HER2 supplied as a hemihydrate for research use. It is orally bioavailable and exhibits antineoplastic activity against EGFR-altered tumors, including exon 20 insertions; intended for laboratory research and not for human use.
  • Hemihydrate form for consistent handling and stability.
  • High purity suitable for biochemical and cellular assays.
  • Brain-penetrant and orally bioavailable properties.
  • Targets EGFR and HER2 alterations, including exon 20 insertions.
  • Supplied in small-mass packs for preclinical research.