Learn More
CEP Enpac Poly-Pail Funnel™ Polyethylene Funnel
SDP
Catalog No. 19087808
Click to view available options
Material:
Polyethylene
Catalog No. Material
19-087808 Polyethylene
1 options
Catalog No. 19-087808 Supplier CEP Supplier No. 3005YE
Polyethylene funnel for spill-free pouring into pails. Fits 3 1/2, 5 and 6 gal. (13, 19, 23L) tight-head pails. Also fits open-top pails with 12 in. diameter (30.5cm). Funnel has 11 in. O.D. (28cm) and 2 3/8 in. high (6cm) sidewall. Spout has 1 1/2 in. O.D. (3.8cm).
Specifications
Material Polyethylene