Learn More
MSC Enpac Spill Pallet, 4-Drum, 66 gal, 6000 lb
Catalog No. 5000782859
Encompass
This spill pallet provides reliable secondary containment for four drums, essential for preventing spills and ensuring compliance in industrial and laboratory environments. Its durable construction and intelligent design facilitate safe handling and storage of hazardous liquids.
  • Forkliftable for ease of transport and repositioning
  • Low profile design allows for convenient drum loading and unloading
  • Integrated drain plug simplifies liquid recovery and cleaning
  • Designed for vertical storage with a 2x2 drum configuration
  • Meets EPA 40 CFR 264.175 regulations for spill containment
Catalog No. 50-007-82859 Supplier MSC Supplier No. 89511695
This spill pallet provides reliable secondary containment for four drums, essential for preventing spills and ensuring compliance in industrial and laboratory environments. Its durable construction and intelligent design facilitate safe handling and storage of hazardous liquids.
  • Forkliftable for ease of transport and repositioning
  • Low profile design allows for convenient drum loading and unloading
  • Integrated drain plug simplifies liquid recovery and cleaning
  • Designed for vertical storage with a 2x2 drum configuration
  • Meets EPA 40 CFR 264.175 regulations for spill containment