Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEP ENPAC™ Spillpal Drum Pallets
SDP

Catalog No. 19048804
Encompass
Encompass_Preferred
Change view
Click to view available options
Dimensions (L x W x H):
2 x 2 x 4 in.
2 x 4 x 4 in.
2 x 8 x 4 in.
4 x 4 x 4 in.
4 x 6 x 4 in.
4 x 8 x 4 in.
Material:
Nylon
Polyethylene
Type:
1-drum Spill Pallet
2-drum Spill Pallet
4-drum In-line Spill Pallet
4-drum Spill Pallet
6-drum Spill Pallet
8-drum Spill Pallet
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H) Material Type
19-048-804 2 x 8 x 4 in. Nylon 4-drum In-line Spill Pallet
19-048-800 2 x 2 x 4 in. Polyethylene 1-drum Spill Pallet
19-048-802 2 x 4 x 4 in. Polyethylene 2-drum Spill Pallet
19-048-803 4 x 4 x 4 in. Nylon 4-drum Spill Pallet
19-048-806 4 x 8 x 4 in. Nylon 8-drum Spill Pallet
19-048-805 4 x 6 x 4 in. Nylon 6-drum Spill Pallet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19-048-804 Supplier CEP Supplier No. 5765YE
Add to Cart
Add to Cart

Provides incidental spill control during temporary storage of drums, machine parts, tanks and generators

  • Flexible four inch foam sidewalls spring back to form, allowing easy drive-in and drive-out with a hand truck or forklift
  • Designed for 37.8L (10 gal.) drums
  • Able to be folded and stored
  • Reusable

Specifications

Certifications/Compliance SPCC, EPA, NPDES
Color Yellow
Capacity (English) 40 gal.
Capacity (Metric) 113.5L
Material Nylon
Type 4-drum In-line Spill Pallet
For Use With (Application) Used to store hazardous material
Holds 4 Drums
Includes Foam sidewalls
Dimensions (L x W x H) 2 x 8 x 4 in.
Length (English) 2 ft.
Length (Metric) 0.6 m
Width (English) 8 ft.
Width (Metric) 2.43 m
Height (English) 4 in.
Height (Metric) 10.16 cm
Weight (English) 5.5 lb.
Weight (Metric) 2.49 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.