Learn More
Medchemexpress LLC HY-134581 5mg , Enpatoran CAS:2101938-42-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1345815MG
Encompass_Preferred
HY-134581 5mg Enpatoran CAS: 2101938-42-3 Enpatoran (M5049) is a potent, orally active and dual TLR7/8 inhibitor with IC50s of 11.1 nM and 24.1 nM in HEK293 cells, respectively. Enpatoran is inactive against TLR3, TLR4 and TLR9. Enpatoran can block molecule synthetic ligands and natural endogenous RNA ligands. Enpatoran exhibits excellent pharmacokinetic properties in vivo. Enpatoran can be used for both innate and adaptive autoimmunity blocking research . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9704