Learn More
Medchemexpress LLC HY-134581A 5mg , Enpatoran (hydrochloride) CAS:2101945-93-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY134581A5MG
Encompass_Preferred
HY-134581A 5mg Enpatoran (hydrochloride) CAS: 2101945-93-9 Enpatoran (M5049) hydrochloride is a potent, orally active and dual TLR7/8 inhibitor with IC50s of 11.1 nM and 24.1 nM in HEK293 cells, respectively. Enpatoran hydrochloride is inactive against TLR3, TLR4 and TLR9. Enpatoran hydrochloride can block molecule synthetic ligands and natural endogenous RNA ligands. Enpatoran hydrochloride exhibits excellent pharmacokinetic properties in vivo. Enpatoran hydrochloride can be used for both innate and adaptive autoimmunity blocking research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0063