Learn More
Aviva Systems Biology Corp ENPEP Antibody - C-terminal region (ARP65562_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65562_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ENPEP. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Human ENPEP. Alternate names: APA, CD249, gp160ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-288-520