Learn More
Medchemexpress LLC Enpp/carbonic anhydrase-IN-1 | 2883495-35-8 | MFCD34676588 | 99.6% | 411.51 g/mol | C23H25NO4S | 10 MG
SDP
Catalog No. 5000287676
Encompass_Preferred
Enpp/carbonic anhydrase-IN-1 is a small-molecule inhibitor that targets ectonucleotide pyrophosphatase/phosphodiesterase (ENPP) family members and carbonic anhydrase isoforms. It has demonstrated antiproliferative activity in cancer cell lines, induces apoptosis, and exhibits low cytotoxicity toward normal cells. The compound is supplied as a solid for research use.
  • Dual-target activity against ENPP family and carbonic anhydrases.
  • Demonstrates antiproliferative activity and induces apoptosis in cancer cells.
  • Low cytotoxicity against normal cells in reported studies.
  • High chemical purity and characterized physicochemical properties.
  • Supplied as a solid with typical solubility in DMSO for assay preparation.
Catalog No. 50-002-87676 Supplier Medchemexpress LLC Supplier No. HY15191610MG
Enpp/carbonic anhydrase-IN-1 is a small-molecule inhibitor that targets ectonucleotide pyrophosphatase/phosphodiesterase (ENPP) family members and carbonic anhydrase isoforms. It has demonstrated antiproliferative activity in cancer cell lines, induces apoptosis, and exhibits low cytotoxicity toward normal cells. The compound is supplied as a solid for research use.
  • Dual-target activity against ENPP family and carbonic anhydrases.
  • Demonstrates antiproliferative activity and induces apoptosis in cancer cells.
  • Low cytotoxicity against normal cells in reported studies.
  • High chemical purity and characterized physicochemical properties.
  • Supplied as a solid with typical solubility in DMSO for assay preparation.