Learn More
Medchemexpress LLC Enpp/carbonic anhydrase-in-2 (compound 1i) | 2883495-39-2 | 99.8% | 429.50 | C23H24FNO4S | 10 MG
SDP
Catalog No. 5000287677
Encompass_Preferred
Enpp/Carbonic anhydrase-IN-2 is a research-grade small-molecule inhibitor active against ENPP family members and carbonic anhydrase isoforms. Reported as compound 1i, it shows antiproliferative activity in cancer cell lines, induces apoptosis, and exhibits low cytotoxicity toward normal cells. Supplied as a purified reference standard for in vitro biochemical and cell-based research assays.
  • Potent inhibition of ENPP enzymes and carbonic anhydrase.
  • Demonstrated antiproliferative activity and induction of apoptosis in cancer cells.
  • Low cytotoxicity against normal cells supports selective activity.
  • High chemical purity suitable for biochemical and cell-based assays.
  • Available in multiple small-scale package sizes for research workflows.
Catalog No. 50-002-87677 Supplier Medchemexpress LLC Supplier No. HY15191710MG
Enpp/Carbonic anhydrase-IN-2 is a research-grade small-molecule inhibitor active against ENPP family members and carbonic anhydrase isoforms. Reported as compound 1i, it shows antiproliferative activity in cancer cell lines, induces apoptosis, and exhibits low cytotoxicity toward normal cells. Supplied as a purified reference standard for in vitro biochemical and cell-based research assays.
  • Potent inhibition of ENPP enzymes and carbonic anhydrase.
  • Demonstrated antiproliferative activity and induction of apoptosis in cancer cells.
  • Low cytotoxicity against normal cells supports selective activity.
  • High chemical purity suitable for biochemical and cell-based assays.
  • Available in multiple small-scale package sizes for research workflows.