Learn More
Cayman Chemical ENPP1 InhIbItr 43 1mg
SDP
Catalog No. 502909665
Encompass_Preferred
An ENPP1 inhibitor (Ki = 41 nM); inhibits tumor growth in an LLC1 murine lung cancer model when administered orally or intravenously at doses of 10 and 100 mg/kg, respectively
Catalog No. 50-290-9665 Supplier Cayman Chemical Supplier No. 368361 MG
An ENPP1 inhibitor (Ki = 41 nM); inhibits tumor growth in an LLC1 murine lung cancer model when administered orally or intravenously at doses of 10 and 100 mg/kg, respectively