Learn More
Cayman Chemical ENPP1 Inhibitor 4e, Purity: ≥98%, 50 mg
SDP
Catalog No. 502916779
Encompass_Preferred
An ENPP1 inhibitor (IC50 = 0.188 µM); inhibits ENPP1 in MDA-MB-231 cells (IC50 = 0.732 µM); selectively cytotoxic to 4T1 murine metastatic breast cancer cells (IC50 = 2.99 µM) over non-cancerous L-02 and 293T cells at 50 µM; increases serum levels of IFN-β in mice when administered at a dose of 0.5 mg/kg in combination with cGAMP
Catalog No. 50-291-6779 Supplier Cayman Chemical Supplier No. 3768750 MG
An ENPP1 inhibitor (IC50 = 0.188 µM); inhibits ENPP1 in MDA-MB-231 cells (IC50 = 0.732 µM); selectively cytotoxic to 4T1 murine metastatic breast cancer cells (IC50 = 2.99 µM) over non-cancerous L-02 and 293T cells at 50 µM; increases serum levels of IFN-β in mice when administered at a dose of 0.5 mg/kg in combination with cGAMP