Learn More
Cayman Chemical ENPP1 InhIbItr C 10mg
SDP
Catalog No. 502978910
Encompass_Preferred
An ENPP1 inhibitor (IC50 = 0.26 µM in a cell-free assay); selective for ENPP1 over ENPP2-7 at 10 µM; decreases ENPP1 activity in MDA-MB-231 and C6 cancer cells at 10 µM
Catalog No. 50-297-8910 Supplier Cayman Chemical Supplier No. 2980910 MG
An ENPP1 inhibitor (IC50 = 0.26 µM in a cell-free assay); selective for ENPP1 over ENPP2-7 at 10 µM; decreases ENPP1 activity in MDA-MB-231 and C6 cancer cells at 10 µM