Learn More
Cayman Chemical ENPP1 Inhibitor C, Purity: ≥98%, CAS: 2378640-92-5, 1 mg
SDP
Catalog No. 502978719
Encompass_Preferred
An ENPP1 inhibitor (IC50 = 0.26 µM in a cell-free assay); selective for ENPP1 over ENPP2-7 at 10 µM; decreases ENPP1 activity in MDA-MB-231 and C6 cancer cells at 10 µM
Catalog No. 50-297-8719 Supplier Cayman Chemical Supplier No. 298091 MG
An ENPP1 inhibitor (IC50 = 0.26 µM in a cell-free assay); selective for ENPP1 over ENPP2-7 at 10 µM; decreases ENPP1 activity in MDA-MB-231 and C6 cancer cells at 10 µM