Learn More
Cayman Chemical ENPP1 Inhibitor C Assay Reagent, 1 ea
SDP
Catalog No. 502894034
Encompass_Preferred
ENPP1 Inhibitor C Assay Reagent has been tested and formulated to work exclusively with Cayman’s ENPP1 Fluorescent Inhibitor Screening Assay Kit (Item No.702090).
Catalog No. 50-289-4034 Supplier Cayman Chemical Supplier No. 7020941 EA
ENPP1 Inhibitor C Assay Reagent has been tested and formulated to work exclusively with Cayman’s ENPP1 Fluorescent Inhibitor Screening Assay Kit (Item No.702090).