Learn More
Cayman Chemical ENPP3 InhIbItr 4g 25mg
SDP
Catalog No. 502916108
Encompass_Preferred
An ENPP1 and ENPP3 inhibitor (IC50s = 0.45 and 0.19 µM, respectively)
Catalog No. 50-291-6108 Supplier Cayman Chemical Supplier No. 3764725 MG
Add to Cart
Add to Cart
An ENPP1 and ENPP3 inhibitor (IC50s = 0.45 and 0.19 µM, respectively)