Learn More
Cayman Chemical ENPP3 Inhibitor 4g, Purity: ≥95%, CAS: 2803505-78-2, 50 mg
SDP
Catalog No. 502918195
Encompass_Preferred
An ENPP1 and ENPP3 inhibitor (IC50s = 0.45 and 0.19 µM, respectively)
Catalog No. 50-291-8195 Supplier Cayman Chemical Supplier No. 3764750 MG
An ENPP1 and ENPP3 inhibitor (IC50s = 0.45 and 0.19 µM, respectively)