Learn More
Cayman Chemical ENPP3 InhIbItr 4t 50mg
SDP
Catalog No. 502915966
Encompass_Preferred
An ENPP3 inhibitor (IC50 = 0.15 µM); selective for ENPP3 over ENPP1 (IC50 = 41.4 µM)
Catalog No. 50-291-5966 Supplier Cayman Chemical Supplier No. 3764650 MG
Add to Cart
Add to Cart
An ENPP3 inhibitor (IC50 = 0.15 µM); selective for ENPP3 over ENPP1 (IC50 = 41.4 µM)