Learn More
Cayman Chemical ENPP3 InhIbItr 5J 10mg
SDP
Catalog No. 502943563
Encompass_Preferred
An ENPP1 and ENPP3 inhibitor (IC50s = 0.59 and 0.62 µM, respectively, for the human enzymes)
Catalog No. 50-294-3563 Supplier Cayman Chemical Supplier No. 4026510 MG
Add to Cart
Add to Cart
An ENPP1 and ENPP3 inhibitor (IC50s = 0.59 and 0.62 µM, respectively, for the human enzymes)