Learn More
Cayman Chemical ENPP3 Inhibitor 5J, Purity: ≥95%, 1 mg
SDP
Catalog No. 502944913
Encompass_Preferred
An ENPP1 and ENPP3 inhibitor (IC50s = 0.59 and 0.62 µM, respectively, for the human enzymes)
Catalog No. 50-294-4913 Supplier Cayman Chemical Supplier No. 402651 MG
An ENPP1 and ENPP3 inhibitor (IC50s = 0.59 and 0.62 µM, respectively, for the human enzymes)