Learn More
Cayman Chemical ENPP3 InhIbItr 5J 5mg
SDP
Catalog No. 502944594
Encompass_Preferred
An ENPP1 and ENPP3 inhibitor (IC50s = 0.59 and 0.62 µM, respectively, for the human enzymes)
Catalog No. 50-294-4594 Supplier Cayman Chemical Supplier No. 402655 MG
An ENPP1 and ENPP3 inhibitor (IC50s = 0.59 and 0.62 µM, respectively, for the human enzymes)