Learn More
Cayman Chemical ENPP3 Inhibitor, Purity: ≥95%, CAS: 333413-02-8, 5 mg
SDP
Catalog No. 502917707
Encompass_Preferred
An ENPP3 inhibitor (Ki = 53.7 nM); selective for ENPP3 over ENPP1, ENPP2, TNAP, CD73, NTPDase 1, NTPDase 2, and NTPDase 3; inhibits CAII and CAIX (Kis = 74.7 and 20.3 nM, respectively)
Catalog No. 50-291-7707 Supplier Cayman Chemical Supplier No. 377605 MG
An ENPP3 inhibitor (Ki = 53.7 nM); selective for ENPP3 over ENPP1, ENPP2, TNAP, CD73, NTPDase 1, NTPDase 2, and NTPDase 3; inhibits CAII and CAIX (Kis = 74.7 and 20.3 nM, respectively)