Learn More
Sino Biological ENPP5 Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 11519T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ENPP5 (rh ENPP5; Catalog#11519-H08H; Q9UJA9; Met1-Gly429). ENPP5 specific IgG was purified by Human ENPP5 affinity chromatography.
Catalog No. 50-224-8638