Learn More
Sino Biological ENPP7 / NPP-7 Antibody, Rabbit MAb 100uL
Supplier:  Sino Biological 10885R001100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human ENPP7 / NPP-7 (rh ENPP7; Catalog#10885-H08H; NP_848638.2; Met 1-Ser 439).
Catalog No. 50-224-7858