Learn More
Sino Biological ENPP7 / NPP-7 Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 10885RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ENPP7 / NPP-7 (rh ENPP7; Catalog#10885-H08H; NP_848638.2; Met 1-Ser 439). ENPP7 specific IgG was purified by human ENPP7 affinity chromatography.
Catalog No. 50-224-8585