Learn More
METTLER TOLEDO pH Sensor InPro 3253SG 120/PT100
Catalog No. NC9368128 Shop All METTLER TOLEDO Products
Encompass
The InPro 3253SG 120/PT100 is a pre-pressurized, liquid filled, low maintenance pH & ORP (Redox) sensor, combined with a PT100 temperature sensor, for in-line measurement in demanding applications. This version is 120 mm long.
Catalog No. NC9368128 Supplier METTLER TOLEDO Supplier No. 52002576
The InPro 3253SG 120/PT100 is a pre-pressurized, liquid filled, low maintenance pH & ORP (Redox) sensor, combined with a PT100 temperature sensor, for in-line measurement in demanding applications. This version is 120 mm long.