Learn More
Cayman Chemical EnramycIn A 10mg
SDP
Catalog No. 502978482
Encompass_Preferred
A lipopeptide antibiotic; inhibits avian myeloblastosis virus reverse transcriptase by 50% at 10 UG/ml in a cell-free assay
Catalog No. 50-297-8482 Supplier Cayman Chemical Supplier No. 4084710 MG
A lipopeptide antibiotic; inhibits avian myeloblastosis virus reverse transcriptase by 50% at 10 UG/ml in a cell-free assay