Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Enramycin, G-Biosciences
SCI_ED

Catalog No. S139782
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139782 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S139782 Supplier G-Biosciences Supplier No. RC1003
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Enramycin is a polypeptide antibiotic that displays strong activity against gram positive bacteria, including methicillin-resistant Staphylococcus aureus (MRSA).

Enramycin is a polypeptide antibiotic that displays strong activity against gram positive bacteria, including methicillin-resistant Staphylococcus aureus (MRSA). It is produced by the fungus Streptomyces fungicidus as Enramycin A (EA) and Enramycin B (EB), with this product containing an approximate 70:30 ratio of EA to EB. Enramycin has been effective against antibiotic resistant bacteria by inhibiting peptidoglycan synthesis for bacterial cell walls

Enramycin has been proposed to halt peptidoglycan synthesis by inhibiting transferase enzyme MurG or by inhibiting transglycosylase activity. Both inhibitory pathways entail enramycin binding to peptidoglycan precursors in a cell wall synthesis pathway. Some studies have shown that enramycin binds the precursor Lipid I and inhibits MurG. Enramycin has also been shown to bind the Lipid II and prevent transglycosylation of the precursor by tranglycosylases.

Chemical Identifiers

Molecular Weight (g/mol) 2355.3

Specifications

Physical Form Powder
Quantity 10 mg
Product Line Gibco
Boiling Point >231°C
Percent Purity >95%
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.