Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Enrasentan | 167256-08-8 | 98.4% | 1 MG
SDP

Catalog No. 5000232996
Encompass_Preferred

Enrasentan is a small-molecule endothelin receptor antagonist used in preclinical research to study endothelin signaling and antihypertensive effects. It is supplied as a characterized solid with documentation for safe handling and storage.

  • Endothelin receptor antagonist used in preclinical pharmacology and in vitro studies.
  • CAS 167256-08-8.
  • Molecular formula C29H30O8; molecular weight 506.54 g/mol.
  • Purity 98.4%.
  • Appearance off-white to pink solid; recommended storage at -20°C for long-term.
  • Documentation available: data sheet, COA, SDS, and handling instructions.

Catalog No. 50-002-32996 Supplier Medchemexpress LLC Supplier No. HY1196671MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Enrasentan is a small-molecule endothelin receptor antagonist used in preclinical research to study endothelin signaling and antihypertensive effects. It is supplied as a characterized solid with documentation for safe handling and storage.

  • Endothelin receptor antagonist used in preclinical pharmacology and in vitro studies.
  • CAS 167256-08-8.
  • Molecular formula C29H30O8; molecular weight 506.54 g/mol.
  • Purity 98.4%.
  • Appearance off-white to pink solid; recommended storage at -20°C for long-term.
  • Documentation available: data sheet, COA, SDS, and handling instructions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.