Learn More
Cole-Parmer Energizer NH22NBP Eveready Rechargeable NiMH Batteries, Size 9V
Catalog No. NC9823775 Shop All Cole-Parmer Products
Encompass
Nickel Metal Hydride (NiMH) batteries provide 70 percent more power than NiCd batteries. They are an ideal alternative to disposable alkaline batteries. The batteries can be recharged and reused up to 1,000 times.
Catalog No. NC9823775 Supplier Cole-Parmer Supplier No. 0950648
Nickel Metal Hydride (NiMH) batteries provide 70 percent more power than NiCd batteries. They are an ideal alternative to disposable alkaline batteries. The batteries can be recharged and reused up to 1,000 times.