Learn More
Cayman Chemical Enrofloxacin, Purity: ≥98%, CAS: 93106-60-6, 10 g
SDP
Catalog No. 502981709
Encompass_Preferred
A fluoroquinolone antibiotic; is active against panels of Campylobacter, E. coli, and Salmonella isolates (mean MIC50s = 0.06, 0.5, and 0.03 µg/ml, respectively); active against equine isolates of L. intracellularis (MICs = 0.125-0.5 µg/ml); inhibits bacterial DNA gyrase; increases survival in a mouse model of enteropathogenic E. coli-induced sepsis at 10 mg/kg
Catalog No. 50-298-1709 Supplier Cayman Chemical Supplier No. 2030110 G
A fluoroquinolone antibiotic; is active against panels of Campylobacter, E. coli, and Salmonella isolates (mean MIC50s = 0.06, 0.5, and 0.03 µg/ml, respectively); active against equine isolates of L. intracellularis (MICs = 0.125-0.5 µg/ml); inhibits bacterial DNA gyrase; increases survival in a mouse model of enteropathogenic E. coli-induced sepsis at 10 mg/kg