Learn More
Apexbio Technology LLC Enrofloxacin 93106-60-6 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063388
Encompass_Preferred
Enrofloxacin a fluoroquinolone antibiotic exerts antibacterial activity by inhibiting bacterial DNA gyrase and topoisomerase IV enzymes essential for DNA replication and transcription This mechanism disrupts the DNA synthesis of pathogenic bacteria resulting in bacteriostatic outcomes In vitro experiments indicate that Enrofloxacin displays antimicrobial activity against Mycoplasma species such as Mycoplasma bovis with a minimum inhibitory concentration (MIC90) around 0 312 g/mL In biomedical research this compound is primarily utilized to establish microbial infection models and to study bacterial resistance mechanisms pharmaceutical efficacy and antibacterial pharmacodynamics
Catalog No. 50-000-63388 Supplier Apexbio Technology LLC Supplier No. B17425.1
Enrofloxacin a fluoroquinolone antibiotic exerts antibacterial activity by inhibiting bacterial DNA gyrase and topoisomerase IV enzymes essential for DNA replication and transcription This mechanism disrupts the DNA synthesis of pathogenic bacteria resulting in bacteriostatic outcomes In vitro experiments indicate that Enrofloxacin displays antimicrobial activity against Mycoplasma species such as Mycoplasma bovis with a minimum inhibitory concentration (MIC90) around 0 312 g/mL In biomedical research this compound is primarily utilized to establish microbial infection models and to study bacterial resistance mechanisms pharmaceutical efficacy and antibacterial pharmacodynamics