Learn More
Bioworld Enrofloxacin, 5 g
Catalog No. 50255955
Encompass_Preferred
Specification:Synonym:1-Cyclopropyl-7-(4-ethyl-1-piperazinyl)-6-Fluoro-1,4-Dihydro-4-Oxo-3-Quinolonecarboxylic Acid; CFPQAn antibiotic that acts against gram-positive and -negative bacteriaHeavy metals: <0.002%Typical working concentration: 5- 25μg/ml Appearance: SolidBoiling Point : 560.46 °C at 760 mmHg (Predicted)Density : 1.39 g/cm3 (Predicted)Melting Point: 218.9-221.1 °CRTECS Number: VB1993650
Catalog No. 50-255-955 Supplier Bioworld Supplier No. 405100011
Specification:Synonym:1-Cyclopropyl-7-(4-ethyl-1-piperazinyl)-6-Fluoro-1,4-Dihydro-4-Oxo-3-Quinolonecarboxylic Acid; CFPQAn antibiotic that acts against gram-positive and -negative bacteriaHeavy metals: <0.002%Typical working concentration: 5- 25μg/ml Appearance: SolidBoiling Point : 560.46 °C at 760 mmHg (Predicted)Density : 1.39 g/cm3 (Predicted)Melting Point: 218.9-221.1 °CRTECS Number: VB1993650