Learn More
Medchemexpress LLC Enrofloxacin-d5 | 1173021-92-5 | 99.8% | 1 MG
SDP
Catalog No. 5000408262
Encompass_Preferred
Enrofloxacin-d5 is the deuterium-labeled version of Enrofloxacin. It is an effective antibiotic with an MIC90 of 0.312 μg/mL for Mycoplasma bovis. Stable heavy isotopes have been incorporated into drug molecules for quantitation during drug development.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium substitution can impact pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-08262 Supplier Medchemexpress LLC Supplier No. HYB0502S1MG
Enrofloxacin-d5 is the deuterium-labeled version of Enrofloxacin. It is an effective antibiotic with an MIC90 of 0.312 μg/mL for Mycoplasma bovis. Stable heavy isotopes have been incorporated into drug molecules for quantitation during drug development.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium substitution can impact pharmacokinetic and metabolic profiles of drugs