Learn More
Medchemexpress LLC Enrofloxacin-d5 | 1173021-92-5 | 99.8% | 5 MG
SDP
Catalog No. 5000409095
Encompass_Preferred
Enrofloxacin-d5 is a deuterium-labeled form of Enrofloxacin, an effective antibiotic with an MIC90 of 0.312 μg/mL for Mycoplasma bovis. Stable heavy isotopes have been incorporated into this drug molecule, primarily as tracers for quantitation during drug development. Deuteration can impact the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium labeling may affect pharmacokinetic and metabolic profiles
Catalog No. 50-004-09095 Supplier Medchemexpress LLC Supplier No. HYB0502S5MG
Enrofloxacin-d5 is a deuterium-labeled form of Enrofloxacin, an effective antibiotic with an MIC90 of 0.312 μg/mL for Mycoplasma bovis. Stable heavy isotopes have been incorporated into this drug molecule, primarily as tracers for quantitation during drug development. Deuteration can impact the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium labeling may affect pharmacokinetic and metabolic profiles