Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Enrofloxacin-d5 | 1173021-92-5 | MFCD28899610 | 99.8% | 364.43 | C19H17D5FN3O3 | 10 MG
SDP

Catalog No. 5000296928
Encompass_Preferred

Enrofloxacin-d5 is the deuterium-labeled isotopologue of the fluoroquinolone antibiotic enrofloxacin, provided as a high-purity analytical reference material for quantitative workflows and method development such as LC-MS/MS.

  • Deuterium-labeled internal standard for LC-MS/MS quantitation.
  • High purity: 99.8%.
  • Molecular weight: 364.43 g/mol.
  • Chemical formula: C19H17D5FN3O3.
  • Supplied as a solid powder; typical pack size 10 MG.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-96928 Supplier Medchemexpress LLC Supplier No. HYB0502S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Enrofloxacin-d5 is the deuterium-labeled isotopologue of the fluoroquinolone antibiotic enrofloxacin, provided as a high-purity analytical reference material for quantitative workflows and method development such as LC-MS/MS.

  • Deuterium-labeled internal standard for LC-MS/MS quantitation.
  • High purity: 99.8%.
  • Molecular weight: 364.43 g/mol.
  • Chemical formula: C19H17D5FN3O3.
  • Supplied as a solid powder; typical pack size 10 MG.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.