Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Enrofloxacin, G-Biosciences
SCI_ED

Catalog No. S139801
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139801 5 g
S139728 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S139801 Supplier G-Biosciences Supplier No. RC1004
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Enrofloxacin is a fluoroquinolone antibiotic targeting gram-positive and gram-negative bacteria as well as some Mycoplasma species.

It has been useful in researching resistance to quinolones in Salmonella enterica, Mycoplasma gallisepticum, and E. coli.

Chemical Identifiers

Molecular Weight (g/mol) 359.39

Specifications

Physical Form Powder
Quantity 5 g
Product Line Gibco
Boiling Point 560.5±50.0°C
Percent Purity 0.98
Grade Molecular Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.