Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Enrofloxacin HCl, G-Biosciences
SCI_ED

Catalog No. S157227
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S157227 100 mg
S139817 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S157227 Supplier G-Biosciences Supplier No. RC1006
Add to Cart
Add to Cart

Enrofloxacin is a fluoroquinolone antibiotic targeting gram-positive and gram-negative bacteria as well as some Mycoplasma species.

It has been useful in researching resistance to quinolones in Salmonella enterica, Mycoplasma gallisepticum, and E. coli.

Chemical Identifiers

Molecular Weight (g/mol) 395.86 g/mol

Specifications

Boiling Point 580.5°C
Quantity 100 mg
Percent Purity >95%
Grade Molecular Biology
Physical Form Powder
Product Line Gibco
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.