Learn More
Medchemexpress LLC HY-B0502A 10g , Enrofloxacin (monohydrochloride) CAS:93106-59-3 Purity:>98%
SDP
Catalog No. 502259400
Encompass_Preferred
HY-B0502A 10g Enrofloxacin (monohydrochloride) CAS: 93106-59-3 Enrofloxacin monohydrochloride (BAY Vp 2674 monohydrochloride) is an effective antibiotic with an MIC90 of 0.312 μg/mL for Mycoplasma bovis. Enrofloxacin monohydrochloride shows inhibitory activity against vaccinia virus (VV). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-225-9400 Supplier Medchemexpress LLC Supplier No. HYB0502A10G
HY-B0502A 10g Enrofloxacin (monohydrochloride) CAS: 93106-59-3 Enrofloxacin monohydrochloride (BAY Vp 2674 monohydrochloride) is an effective antibiotic with an MIC90 of 0.312 μg/mL for Mycoplasma bovis. Enrofloxacin monohydrochloride shows inhibitory activity against vaccinia virus (VV). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.