Learn More
Medchemexpress LLC Enrofloxacin (monohydrochloride) | 93106-59-3 | 99.7% | 500 MG
SDP
Catalog No. 501970112
Encompass_Preferred
Enrofloxacin monohydrochloride is an effective antibiotic with an MIC90 of 0.312 μg/mL for Mycoplasma bovis. It also exhibits inhibitory activity against vaccinia virus (VV). This product is for research use only.
  • Effective antibiotic
  • Exhibits inhibitory activity against vaccinia virus (VV)
  • Shows an MIC90 of 0.312 μg/mL for Mycoplasma bovis
  • Stable under recommended storage conditions
Catalog No. 50-197-0112 Supplier Medchemexpress LLC Supplier No. HYB0502A500MG
Enrofloxacin monohydrochloride is an effective antibiotic with an MIC90 of 0.312 μg/mL for Mycoplasma bovis. It also exhibits inhibitory activity against vaccinia virus (VV). This product is for research use only.
  • Effective antibiotic
  • Exhibits inhibitory activity against vaccinia virus (VV)
  • Shows an MIC90 of 0.312 μg/mL for Mycoplasma bovis
  • Stable under recommended storage conditions