Learn More
Medchemexpress LLC Enrofloxacin monohydrochloride | 93106-59-3 | 99.73% | 1 ML
SDP
Catalog No. 5000407996
Encompass_Preferred
Enrofloxacin monohydrochloride is an effective antibiotic (MIC90 of 0.312 μg/mL for Mycoplasma bovis) that also exhibits inhibitory activity against vaccinia virus (VV). It is provided as a white to off-white solid with a molecular weight of 395.86.
  • Effective antibiotic
  • Inhibitory activity against vaccinia virus (VV)
  • High purity of 99.73%
  • Provided as a solid
Catalog No. 50-004-07996 Supplier Medchemexpress LLC Supplier No. HYB0502A10MM/1ML
Enrofloxacin monohydrochloride is an effective antibiotic (MIC90 of 0.312 μg/mL for Mycoplasma bovis) that also exhibits inhibitory activity against vaccinia virus (VV). It is provided as a white to off-white solid with a molecular weight of 395.86.
  • Effective antibiotic
  • Inhibitory activity against vaccinia virus (VV)
  • High purity of 99.73%
  • Provided as a solid