Learn More
Aviva Systems Biology Corp ENSA Antibody - N-terminal region (ARP53514_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53514_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ENSA. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ENSA. Alternate names: MGC4319, MGC78563, MGC8394, ARPP-19eApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-844