Learn More
Medchemexpress LLC Ensartinib dihydroch 100mg | 2137030-98-7 | 100MG
SDP
Catalog No. 5000241857
Encompass_Preferred
Ensartinib dihydrochloride (X-396 dihydrochloride) is a potent and dual ALK/MET inhibitor with IC50s of 0 4 nM and 0 74 nM respectively
Catalog No. 50-002-41857 Supplier Medchemexpress LLC Supplier No. HY103714A100MG
Ensartinib dihydrochloride (X-396 dihydrochloride) is a potent and dual ALK/MET inhibitor with IC50s of 0 4 nM and 0 74 nM respectively