Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ensartinib dihydrochloride | 2137030-98-7 | 99.7% | 634.36 | C26H29Cl4FN6O3 | 10 MG
SDP

Catalog No. 5000243287
Encompass_Preferred

Ensartinib dihydrochloride is the dihydrochloride salt of ensartinib (X-396), a potent small-molecule inhibitor targeting anaplastic lymphoma kinase (ALK) and MET for research use. It is provided as a high-purity solid for in vitro and preclinical studies and is not intended for human use.

  • Potent dual ALK/MET inhibitor with IC50s of <0.4 nM (ALK) and 0.74 nM (MET).
  • High purity (99.73%).
  • Molecular weight 634.36 g/mol.
  • Molecular formula C26H29Cl4FN6O3.
  • Solid, white to light yellow powder.
  • Supplied as a 10 mg research quantity.
  • Store sealed, away from moisture; in solvent: -80°C for 6 months, -20°C for 1 month.

Catalog No. 50-002-43287 Supplier Medchemexpress LLC Supplier No. HY103714A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ensartinib dihydrochloride is the dihydrochloride salt of ensartinib (X-396), a potent small-molecule inhibitor targeting anaplastic lymphoma kinase (ALK) and MET for research use. It is provided as a high-purity solid for in vitro and preclinical studies and is not intended for human use.

  • Potent dual ALK/MET inhibitor with IC50s of <0.4 nM (ALK) and 0.74 nM (MET).
  • High purity (99.73%).
  • Molecular weight 634.36 g/mol.
  • Molecular formula C26H29Cl4FN6O3.
  • Solid, white to light yellow powder.
  • Supplied as a 10 mg research quantity.
  • Store sealed, away from moisture; in solvent: -80°C for 6 months, -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.