Learn More
Medchemexpress LLC Ensartinib dihydrochloride | 2137030-98-7 | MFCD31657344 | 99.7% | 634.36 g/mol | C26H29Cl4FN6O3 | 1 ML
SDP
Catalog No. 5000244128
Encompass_Preferred
Ensartinib dihydrochloride is the dihydrochloride salt of ensartinib, a potent dual ALK and MET tyrosine kinase inhibitor used in preclinical research to investigate ALK-driven cancers and resistance mutations. It is supplied in high-purity solid and solution formats for laboratory use.
  • Potent dual ALK and MET inhibition with sub-nanomolar IC50s.
  • High purity (≈99.7%) suitable for analytical and in vitro use.
  • Available as a 10 mM solution (1 mL) and multiple solid pack sizes.
  • Soluble in DMSO for convenient stock preparation.
  • Intended for preclinical kinase profiling and resistance mechanism studies.
Catalog No. 50-002-44128 Supplier Medchemexpress LLC Supplier No. HY103714A10MM 1ML
Ensartinib dihydrochloride is the dihydrochloride salt of ensartinib, a potent dual ALK and MET tyrosine kinase inhibitor used in preclinical research to investigate ALK-driven cancers and resistance mutations. It is supplied in high-purity solid and solution formats for laboratory use.
  • Potent dual ALK and MET inhibition with sub-nanomolar IC50s.
  • High purity (≈99.7%) suitable for analytical and in vitro use.
  • Available as a 10 mM solution (1 mL) and multiple solid pack sizes.
  • Soluble in DMSO for convenient stock preparation.
  • Intended for preclinical kinase profiling and resistance mechanism studies.