Learn More
Mopec Ensemble Dual Monitor Mount Arm
SDP
Catalog No. 23987038 Shop All Mopec Products
Change view
Click to view available options
For Use With (Equipment):
Dual Monitor Mount
Catalog No. For Use With (Equipment)
23-987-038 Dual Monitor Mount
1 options
Catalog No. 23-987-038 Supplier Mopec Supplier No. AX142
Add to Cart
Add to Cart
Designed to mount two monitors into the existing track on the column opposite the sink
  • Designed to mount two monitors into the existing track on the column opposite the sink
  • Once installed, track allows arm to be moved up and down the track by loosening two large, knurled knobs, without needing any tools
  • Keyboard arm components are powder-coated aluminum and steel for the brackets
  • Has several pivot points to articulate up and down, and side to side.
  • Can be pulled out when in use, then pushed back when not in use
  • Fits all Mopec Maestro and Maestro Encore grossing stations
Specifications
Product Type Grossing Station
For Use With (Equipment) Dual Monitor Mount