Description
Construction
- Designed to be an optional additional grid plate for your grossing station
Materials
- 304 stainless steel construction with a polished No. 4 finish
Dimensions
- 11.9L x 18.3D x 0.8H in.
Capacity
- 40 lb.
Compatibility
- Fits all Mopec Maestro and Maestro Encore grossing stations
Specifications
Specifications
| Product Type | Grid Plate |
| Includes | Grid Plate |