Learn More
Mopec™ Ensemble Lower Surface Light Bar
SDP
Catalog No. 22011110 Shop All Mopec Products
Change view
Click to view available options
For Use With (Equipment):
Mopec Maestro and Maestro Encore Grossing Stations
Catalog No. For Use With (Equipment)
22-011-110 Mopec Maestro and Maestro Encore Grossing Stations
1 options
Catalog No. 22-011-110 Supplier Mopec™ Supplier No. AX049
Add to Cart
Add to Cart
Designed to mount under the shelf/above the vent grille on the Maestro Encore Grossing station.
NOTE: This option must be ordered and at the time of purchase and installed during manufacturing of the grossing station. It cannot be installed in the field after delivery.
Specifications
Product Type Lower Surface Light Bar
Material Painted Metal frame with a polycarbonate lens, LED light diodes
Description Daylight white light, 6500K
For Use With (Equipment) Mopec Maestro and Maestro Encore Grossing Stations